missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Milli-Q™ Ultrapure Water Systems Accessory, Cartridges

Catalog No. QGARD00D2
Change view
Click to view available options
Type:
Polishing Cartridge
Polishing Cartridge and Filter
Purification Cartridge
Q-Gard 2 Purification Pack
Replacement Cartridge
Ultrafiltration Cartridge
Model:
Pyrogard™ 5000
Q-Gard 1
Q-Gard 2
Q-Gard™ Boron
Quantum EX
Quantum EX (Organex)1
Quantum ICP
Quantum IX
Quantum IX (Ionex)1
Quantum VX
Purification Method:
Ion Exchange
Ion Exchange, Activated Carbon
Ion Exchange, Activated Carbon, Depth Filtration
Ion Exchange, Activated Carbon, Membrane Filter
Ion Exchange, Membrane Filter
Ultrafiltration
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Model Purification Method
QGARD00D2 Q-Gard 2 Purification Pack Q-Gard 2 Ion Exchange, Activated Carbon, Depth Filtration
QTUM00ICP Polishing Cartridge Quantum ICP Ion Exchange
QTUMMPEEX Polishing Cartridge and Filter Quantum EX (Organex)1 Ion Exchange, Activated Carbon, Membrane Filter
QTUMMPEIX Polishing Cartridge and Filter Quantum IX (Ionex)1 Ion Exchange, Membrane Filter
QTUM000IX Polishing Cartridge Quantum IX Ion Exchange
QGARD00R1 Replacement Cartridge Q-Gard 1 Ion Exchange, Activated Carbon
CDUFHF05K Ultrafiltration Cartridge Pyrogard™ 5000 Ultrafiltration
QGARD00B1 Purification Cartridge Q-Gard™ Boron Ion Exchange
QTUM000EX Polishing Cartridge Quantum EX Ion Exchange, Activated Carbon
QTUM000VX Polishing Cartridge Quantum VX Ion Exchange, Activated Carbon
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. QGARD00D2 Supplier MilliporeSigma™ Supplier No. QGARD00D2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Choose from a wide selection of replacement cartridges

Filter Types

  • Activated carbon
  • Ion exchange
  • Membrane

Specifications

Feedwater Source Deionized
Purification Method Ion Exchange, Activated Carbon, Depth Filtration
For Use With (Equipment) Milli-Q Systems
Model Q-Gard 2
Type Q-Gard 2 Purification Pack