missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Milli-Q™ Advantage A10 Ultrapure Water Purification System Accessory, Quantum Polishing Cartridges

Catalog No. QTUM0TEX1
Change view
Click to view available options
Type:
Quantum TEX Cartridge Filter
Quantum TIX Cartridge Filter
Resin Type:
Ionex
Organex and Ionex
Purification Method:
Ion Exchange
Ion Exchange, Activated Carbon
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Resin Type Purification Method
QTUM0TEX1 Quantum TEX Cartridge Filter Organex and Ionex Ion Exchange, Activated Carbon
QTUM0TIX1 Quantum TIX Cartridge Filter Ionex Ion Exchange
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. QTUM0TEX1 Supplier MilliporeSigma™ Supplier No. QTUM0TEX1
Only null left

Quantum polishing catridge removes ionic and organic contaminants below trace levels to match required water quality requirements

Quantum™ TEX Features

  • Contains Ionex resins for trace ion levels
  • Feedwater nature: Ultrapure
  • Technology: Activated carbon, Ion exchange
Quantum™ TIX Features
  • Contains Organex and Ionex resins for trace levels of organics and ions
  • Feedwater nature: Ultrapure
  • Technology: Activated carbon, Ion exchange

Spectrophotometry, spectroscopy, chromatography, and biochemistry

Certifications

ISO 9001 v.2000 and ISO 14001

Compliance

USP 645 suitability requirements

Specifications

Purification Method Ion Exchange, Activated Carbon
Resin Type Organex and Ionex
For Use With (Application) Spectrophotometry, spectroscopy, chromatography, and biochemistry
For Use With (Equipment) Milli-Q™ Systems
Type Quantum TEX Cartridge Filter