missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MCT4 Control Peptide

Catalog No. AG201MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG201MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG201MI Supplier MilliporeSigma™ Supplier No. AG201
Only null left

Specifications

Gene ID (Entrez) NM_001042422.1, NM_001042423.1, NM_004207.2
Concentration 1 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation Liquid in PBS containing 0.1% BSA and 0.1% sodium azide.
Gene Symbol SLC16A3, MCT3, MGC138472, MGC138474, MCT4
Product Type MCT4, control peptide for AB3550P
Quantity 100 μg
Regulatory Status RUO - research use only