missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Magna GrIP™ Rack

Catalog No. 20400
Change view
Click to view available options
Positions:
8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Positions
20400 8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options

Strong magnets can cause serious injury to people with Pacemakers or Automatic Internal Cardiac Defibrillators. Strong magnets can attract tools and metal objects, causing pinch, cut or entrapment hazards. Keep tools, metal objects, and magnetic media away from magnets. Do not store near computers, cell phones, or other electronic equipment.

Catalog No. 20400 Supplier MilliporeSigma™ Supplier No. 20400

Strong magnets can cause serious injury to people with Pacemakers or Automatic Internal Cardiac Defibrillators. Strong magnets can attract tools and metal objects, causing pinch, cut or entrapment hazards. Keep tools, metal objects, and magnetic media away from magnets. Do not store near computers, cell phones, or other electronic equipment.

Designed to rapidly and easily pellet samples bound to magnetic particles from the supernatant.

  • In its horizontal position, the rack contains 8 holes suitable for 1.5 to 2.0mL tubes or spin columns
  • May also be used with either 15 or 0.5mL tubes simply by standing the rack on either side
  • Holes in the rack are slightly angled to maximize exposure to the magnets and to ease removal of the liquid from the tube
  • Contains 4 neodinium magnets used for ChIP and RIP assays
  • Polyethylene

Store at room temperature. Do not autoclave. If necessary, wash in soapy water, rinse, and air dry. The rack is compatible with alcohols; avoid exposure to organic solvents including benzene, toluene, xylene, chloroform, or acetone.

Specifications

Autoclavable Not Autoclavable
Color White
Holds Microtubes (1.5 to 2 mL)
Material Polyethylene
Positions 8
Shape Rectangular
Sterility Non-sterile
For Use With (Application) ChIP and RIP assays
Product Type Magnetic Rack
Array 4 x 2 or 2 x 4