missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ m-ColiBlue24™ Broth Culture Media

Catalog No. M00PMCB24
Change view
Click to view available options
Quantity:
50 x 2 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M00PMCB24 50 x 2 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M00PMCB24 Supplier MilliporeSigma™ Supplier No. M00PMCB24
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Simultaneously detects total coliforms and E. coli in water and beverages using standard membrane filtration equipment

  • Ready-to-use medium comes prepared and packaged in single-test 2mL plastic ampules
  • Combination of enzymatic indicators in the medium differentiates E. coli, which form blue colonies, from non-E. coli coliforms, which form red colonies

Order Info

Prepared and packaged in single-test 2mL plastic ampules.

Specifications

Appearance Blue Liquid
Broth Type m-ColiBlue24 Broth
Certifications/Compliance EPA Approved
Color Blue
Content And Storage 2°C to 8°C
Additive Enzymatic Indicator, Special Inhibitors
Detects Total Coliforms and Escherichia coli
Form Liquid
Format Ready-to-use Liquid, Ampule
Manufacturing Origin United States
Media Type m-ColiBlue24 Broth
Product Type Microbiology Media
Sterility Sterile
Test Type Water Monitoring
Fill Volume 2 mL
Formulation Enzymatic Indicator and Special Inhibitors
For Use With (Application) For testing drinking Water and ambient Water, Total Coliform and Escherichia coli detection
Incubation Conditions 35°C ±0.5°C
Incubation Time 24 hr.
No. per Pack 50
Packaging Type Ampule
Quantity 50 x 2 mL
Show More Show Less

Is a patented product of Hach Company, U.S. Patents 5,650,290 and 5,849,515.