Learn More
MilliporeSigma™ LiChrolut™ Sorbents
Constant batch-to-batch quality ensured with quality control of raw materials
Supplier:  MilliporeSigma™ 1.19687.0001
 View more versions of this product
Catalog No. M1196870001
Only null left
Description
Description
  • The specially developed, validated and certificated production processes ensure a high degree of purity
  • The multi-stage purification procedures carried out on LiChrolut raw materials become particularly valuable when it comes to trace analysis
Specifications
Chemical Identifiers
SPE Sorbent
Polypropylene (Tube)
Approx. 0.600 m2/g
Specifications
Filling amount: 500 mg
LiChrolut RP-18
Polypropylene (Tube)
Approx. 0.600 m2/g
40 to 63 μm
SPE Sorbent
30/Pk.
6 mL
Product Suggestions
Product Suggestions