Learn More
MilliporeSigma™ LiChrolut™ Extraction Unit
for sample preparation LiChrolut™
Supplier: MilliporeSigma™ 1198510001
Description
All the individual steps associated with solid-phase extraction can be carried out using the LiChrolut™ extraction unit rapidly and reliability.This transparent and vacuum â suitable unit made of glass can be used to prepare up to 12 samples simultaneously.Specifications
| Extraction Unit, Complete |