missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Individual Cartridges for Milli-Q™ and Milli-RO™ Bowl-Style Systems

Catalog No. CPMB01202
Change view
Click to view available options
Model:
Ion-Ex Cartridge
Organex-Q cartridge
Super-C Carbon Cartridge
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
CPMB01202 Ion-Ex Cartridge
CDFC01204 Super-C Carbon Cartridge
CDEX01201 Organex-Q cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CPMB01202 Supplier MilliporeSigma™ Supplier No. CPMB01202

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For Milli-Q

For Milli-Q™ Bowl systems

Specifications

Purification Method Depth Filtration
Filter Size 12 in.
For Use With (Equipment) Milli-Q™ and Milli-RO™ bowl-style systems
Includes 12 in. (30cm) cartridge removes ionized impurities from water pretreated by reverse osmosis, deionization or distillation. Constructed of pure polypropylene with thermally bonded endcaps to reduce the risk of extractables contamination.
Model Ion-Ex Cartridge
Type Replacement Cartridge