missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Individual Cartridges for Milli-Q™ and Milli-RO™ Bowl-Style Systems

Catalog No. CDFC01204
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Model:
Ion-Ex Cartridge
Organex-Q cartridge
Super-C Carbon Cartridge
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Model
CDFC01204 Super-C Carbon Cartridge
CDEX01201 Organex-Q cartridge
CPMB01202 Ion-Ex Cartridge
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Catalog No. CDFC01204 missing translation for 'mfr' MilliporeSigma™ missing translation for 'supplierNo' CDFC01204

For Milli-Q

For Milli-Q Bowl systems

Specifications

Purification Method Activated Carbon
Filter Size 12 in.
For Use With (Equipment) Milli-Q™ and Milli-RO™ bowl-style systems
Includes 12 in. (30cm) cartridge removes organics from water pretreated by reverse osmosis, deionization or distillation. Use in combination with prefilter CDPR M12 06.
Model Super-C Carbon Cartridge
Type Replacement Cartridge