missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Human WNT-1 Recombinant Protein

Catalog No. GF175
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF175 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF175 Supplier MilliporeSigma™ Supplier No. GF175
Only null left

Specifications

Name Human WNT-1 Recombinant Protein
Quantity 10 μg
Source E. coli
Cross Reactivity Mouse
Recombinant Recombinant
Purity or Quality Grade Endotoxin free and greater than 98% pure by SDS-PAGE gel and HPLC analyses