missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Human SDF-1 alpha (CXCL12) Recombinant Protein, Animal Free

Catalog No. GF344
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF344 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF344 Supplier MilliporeSigma™ Supplier No. GF344
Only null left

Specifications

Name Human SDF-1 alpha (CXCL12) Recombinant Protein
Quantity 10 μg
Source E. coli
Recombinant Recombinant
Purity or Quality Grade Endotoxin level is less than 0.1ng/μg of protein