missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ , Human Prostate Epithelial Cells, Adult

Catalog No. SCCE019
Change view
Click to view available options
Quantity:
500,000 cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCCE019 500,000 cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCCE019 Supplier MilliporeSigma™ Supplier No. SCCE019

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

provides an ideal serum-free culture model, without retinoic acid, for the study of prostate cells when grown in our recommended medium.

Millipore’s Prostate Cell line, when grown in our recommended medium, provides an ideal serum-free culture model, without retinoic acid, for the study of prostate cells. Common uses for these cells are studies of hormonal regulation of the prostate, regulation and control of the secretory function of prostate cells and as a control for the study of prostate cancer. These cells are not exposed to antimicrobials or phenol red when cultured in EpiGRO™ Prostate Complete Media Kit. Prostate Cells are cryopreserved at passage 2 to ensure highest viability and plating efficiency.

Specifications

For Use With (Application) Cell Culture
Quantity 500,000 cells
Source Human Prostrate