missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Human MIF Recombinant Protein
Specifications
Specifications
| Name | Human MIF Recombinant Protein |
| Quantity | 25 μg |
| Source | Insect cells |
| Recombinant | Recombinant |
| Purity or Quality Grade | Endotoxin level is less than 0.1ng/μg of protein |