missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Human ES/iPS Neural Differentiation Medium

Catalog No. scm111
Change view
Click to view available options
Quantity:
100 mL/Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM111 100 mL/Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM111 Supplier MilliporeSigma™ Supplier No. SCM111
Only null left

a serum free/chemically defined optimized media for the rapid production of terminally differentiated neurons from human neural progenitor cells.

Specifications

For Use With (Application) Cell Differentiation, Neuroscience
Quantity 100 mL/Kit
Content And Storage -20°C