missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Human BDNF Recombinant Protein, Animal Free

Catalog No. gf301
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
GF301 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF301 Supplier MilliporeSigma™ Supplier No. GF301
Only null left

Specifications

Name Human BDNF Recombinant Protein, Animal Free
Quantity 10 μg
Source E. coli
Cross Reactivity Chicken, Frog, Mouse, Rat, Hamster, Guinea Pig, Monkey
Recombinant Recombinant
Purity or Quality Grade Endotoxin free and greater than 98% pure by SDS-PAGE gel and HPLC analyses