missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Human Activin A Recombinant Protein, Animal Free
Specifications
Specifications
| Name | Human Activin A Recombinant Protein, Animal Free |
| Quantity | 10 μg |
| Source | E. coli |
| Cross Reactivity | Chicken, Dog, Frog, Mouse, Human, Mouse |
| Recombinant | Recombinant |
| Purity or Quality Grade | Endotoxin free and greater than 97% pure by SDS-PAGE gel and HPLC analyses |