Learn More
MilliporeSigma™ HPTLC Cellulose Plates, Glass Backed
Glass HPTLC Cellulose plates.
Supplier: MilliporeSigma™ 1150350001
Description
Cellulose plates are used to analyze polar substances. An organic sorbent, cellulose is perfect for separating hydrophilic substances by partition chromatography. Typical applications include the analysis of amino acids, carbohydrates, and phosphates as well as nucleic acid and nucleic acid derivatives. We offer cellulose plates in two grades: TLC for conventional and HPTLC for demanding, high-performance separations.Specifications
| Glass | |
| 25/Pk. | |
| 254 and 366 nm | |
| Cellulose F | |
| 10 x 10 cm | |
| 10 cm |
| 60 Å | |
| Cellulose HPTLC Plate | |
| Analysis of Polar Substances, Separation of Hydrophilic Substances by Partition Chromatography, for Investigating in-Vitro Samples Derived From The Human Body Without Any Medical Objective | |
| 200 μm | |
| 10 cm |