missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Histamine Receptor 4 Control Peptide
Specifications
Specifications
| Gene ID (Entrez) | NM_021624.2 |
| Concentration | 1.0 mg/mL |
| Content And Storage | -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles |
| Format | Liquid |
| Formulation | Liquid in PBS containing 0.1% sodium azide. |
| Gene Symbol | HRH4, MGC133027, GPCR105, HH4R, GPRv53, H4R, H4, AXOR35, SP9144, BG26 |
| Product Type | Histamine Receptor 4, control peptide for AB5663P |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |