missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ GDF-8/Myostatin, Human, Mouse, Rat, Recombinant Protein

Catalog No. 34576210UG
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34576210UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34576210UG Supplier MilliporeSigma™ Supplier No. 34576210UG
Only null left
Recombinant, human mouse rat growth differentiation factor 8 (GDF-8) consisting of amino acids 268-376 and expressed in NS0-derived murine myeloma cells. GDF-8 is expressed in developing and adult skeletal muscle and is 100% identical to mature human, rat, mouse, and cow at the amino acid sequence level. Useful for the inhibition of preadipocyte differentiation to adipocytes.

Specifications

Formulation Lyophilized from 0.2μM filtered Acetonitrile, TFA, BSA.
Preparation Method Reconstitute to 100μg/mL in sterile 4mM HCl containing at least 0.1% human or bovine serum albumin.
Purification Method SDS-PAGE
Quantity 10 μg
Immunogen GDF-8/Myostatin
Storage Requirements Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Regulatory Status RUO
Species Human, Mouse, Rat
Recombinant Recombinant
Form Solid
Purity or Quality Grade ≥80%
Show More Show Less