missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Gastrin I, Human peptide hormone
Description
Synthetic, human gastrin I. Peptide hormone produced in the stomach. Responsible for stimulation of gastric acid secretion. Delays gastric emptying and increases blood flow in the gastric mucosa.
Specifications
Specifications
| Species | Human |
| Purification Method | HPLC |
| Purity | ≥98% |
| Color | White |
| Content And Storage | Following reconstitution, aliquot and freeze (−20°C). Stock solutions are stable for up to 2 months at −20°C. |
| Format | Solid |
| Formulation | Supplied as a trifluoroacetate salt. Sold on the basis of peptide content. |
| Product Type | Gastrin I |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |