missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ GABA B Receptor R1 Control Peptide

Catalog No. AG324MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG324MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG324MI Supplier MilliporeSigma™ Supplier No. AG324
Only null left

Specifications

Gene ID (Entrez) NM_001470.1, NM_021903.1, NM_021904.1, NM_021905.1
Concentration 1.0 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation Liquid in PBS.
Gene Symbol GABBR1, dJ271M21.1.1, GABBR1-3, GABAB(1e), hGB1a, GABABR1, dJ271M21.1.2, Gb1, GABA-B-R1, GPRC3A
Product Type GABA B Receptor R1, control peptide for AB1531
Quantity 100 μg
Regulatory Status RUO - research use only