Learn More
MilliporeSigma™ EXtrelut™ NT Prepacked Columns
Catalog No. M1150960001
Change view
Click to view available options
Type:
EXtrelut NT 1 Prepacked Column, Glass
EXtrelut NT 20 Prepacked Column, Polyethylene
EXtrelut NT 3 Prepacked Column, Glass
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
M1150960001 EXtrelut NT 20 Prepacked Column, Polyethylene
M1150940001 EXtrelut NT 1 Prepacked Column, Glass
M1150950001 EXtrelut NT 3 Prepacked Column, Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1150960001 Supplier MilliporeSigma™ Supplier No. 1.15096.0001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
pre-packed columns for extraction of lipophilic compounds from aqueous solutions
Extrelut™ NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut™ NT1 can take up a maximum of one ml, Extrelut™ NT3 threemL and Extrelut™ NT20 up to twentymL of sample.
Specifications
For Use With (Application) To Obtain High Degree Purity
Sorbent SPE Sorbent
Type EXtrelut NT 20 Prepacked Column, Polyethylene