Learn More
MilliporeSigma™ EXtrelut™ NT Prepacked Columns
pre-packed columns for extraction of lipophilic compounds from aqueous solutions
Supplier:  MilliporeSigma™ 1.15095.0001
 View more versions of this product
Catalog No. M1150950001
Only null left
Description
Description
Extrelut™ NT simplifies liquid-liquid extraction in its most effective form by using a chemically inert, wide-pore and highly pure diatomaceous earth based solid phase. Depending on the sample volume three types of columns are available: Extrelut™ NT1 can take up a maximum of one ml, Extrelut™ NT3 threemL and Extrelut™ NT20 up to twentymL of sample.
Specifications
Chemical Identifiers
To Obtain High Degree Purity
SPE Sorbent
Specifications
To Obtain High Degree Purity
SPE Sorbent
EXtrelut NT 3 Prepacked Column, Glass
Product Suggestions
Product Suggestions