missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ ESGRO™-2i Selective Medium Kit

Catalog No. p-5235539
Change view
Click to view available options
Quantity:
100 mL
200 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SF016100 100 mL
SF016200 200 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SF016100 Supplier MilliporeSigma™ Supplier No. SF016100
Only null left

provided with a selective GSK3β and Mek 1/2 inhibitors to enhance viability of mouse embryonic stem and iPS cells in the absence of serum and feeder cells. Supports a naive/ground pluripotent state.

Two stages of mouse pluripotency have been stabilized to date through in vitro culture. ‘Naive’ represents the earliest stage, which can be enriched for a purer population, termed ‘ground state’. Naive most closely approximates embryonic epiblast in the preimplantation blastocyst. The primed state most closely approximates in vivo immediate postimplantation epiblast. There are clear differences between these stages in culture, expression, epigenetics (including appropriate X-inactivation) and developmental competence. The fact that naive mESCs can generate whole animals through tetraploid complementation while primed mESCs cannot, is a particularly compelling reason for heightened expectations from naive hESCs.

Specifications

For Use With (Application) Stem Cell Culture
Quantity 100 mL
Content And Storage Store in dark at -20° C