missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Epithelial Sodium Channel γ Control Peptide

Catalog No. AG340MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG340MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG340MI Supplier MilliporeSigma™ Supplier No. AG340

Specifications

Gene ID (Entrez) NM_001039.3
Concentration 1 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation Liquid in PBS pH 7.4, containing 0.1% sodium azide.
Gene Symbol SCNN1G, SCNEG, Gamma-ENaC, ENaCg, Gamma-NaCH, ENaCgamma, PHA1
Product Type Epithelial Sodium Channel γ, control peptide for AB3534P
Quantity 100 μg
Regulatory Status RUO - research use only