missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EmbryoMax™ DMEM (1X), w/4.5g/L Glucose, 2.25g/L Sodium Bicarbonate, and L-Glutamine

Catalog No. SLM120B
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SLM120B 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SLM120B Supplier MilliporeSigma™ Supplier No. SLM120B

Used for routine mouse embryonic stem cell culture applications.

  • Format: Liquid

Specifications

Concentration 1 X
Form Liquid
Product Type DMEM Medium
With Additives Glucose, Sodium Bicarbonate, Glutamine
For Use With (Application) Stem cell culture
Quantity 500 mL
Content And Storage 2-8°C