missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Embryoid Body (EB) Formation Medium

Catalog No. SCM018
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM018 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM018 Supplier MilliporeSigma™ Supplier No. SCM018

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

optimized and qualified to support the formation of embryoid body.

The Embryoid Body (EB) Formation Medium has been optimized and qualified to support the formation of embryoid body. The medium can be used to form EB from hanging drops or suspension culture on non-adhesive Petri dishes. It has been validated for the differentiation of mouse ES cells into neural, adipogenic and cardiomyogenic cell lineages.

Specifications

For Use With (Application) Cell Differentiation
Quantity 100 mL
Content And Storage -20°C