missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Elix™ Essential 3, 5, 10, 15 Water Purification System
Choose a Millipore Elix Essential Water Purification System for your lab to ensure consistently-pure, reliable water quality for optimal results.
Supplier: MilliporeSigma™ ZLXE0150US
Description
Ideal for laboratories needing an easy-to-use economical solution that provides a constant and reliable source of Type 2 pure water.
- Patented Elix™ module, which uses electricity (instead of harsh chemicals and water) to continuously regenerate the resins and remove ions from tap water
- Intuitive system controls, with system alert and alarm icons shown on a color-coded backlit LCD display
- High-precision monitoring equipment to ensure precise and reliable measurement of key water quality parameters
- RFID technology for traceability of Progard™ pretreatment packs
- New ergonomic pack locking system to make Progard™ pack changes a snap
- Small system footprint, allowing multiple installation possibilities
- Has resistivity values greater than 5 Mohm.cm at 25°C and less than 30ppb TOC, exceeding the quality of double distilled water
Feed to laboratory equipment, Preparation of microbiological media, buffer and pH solutions, Histology, Chemical reactions run in water, Manual glassware rinsing
Specifications
| <10 CFU/mL | |
| 15 L/hr. | |
| >5 Mohm.cm | |
| Elix Essential Water Purification System | |
| 87 psi (6 bar) | |
| Without UV lamp | |
| 26.8 cm | |
| 47 cm | |
| 33.9 cm | |
| 12.3 to 14 kg | |
| 100/230 V |
| Tap | |
| Type 2 | |
| 30 ppb | |
| Color Graphic Display | |
| Feed to laboratory equipment, preparation of microbiological media, buffer and pH solutions, histology, chemical reactions run in water, manual Glassware rinsing | |
| 10.55 in. | |
| 18.5 in. | |
| 13.35 in. | |
| 32.95 to 37.51 lb. | |
| 100/230 V 50/60 Hz | |
| 50/60 Hz |