missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Dopamine D4 Mouse Control Peptide
Specifications
Specifications
| Gene ID (Entrez) | NM_000797.2 |
| Concentration | 1.0 mg/mL |
| Content And Storage | -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. |
| Format | Liquid |
| Formulation | Liquid in PBS, pH 7.4 containing 0.02% sodium merthiolate. |
| Gene Symbol | DRD4, D, D4DR |
| Product Type | Dopamine D4, Mouse, Ms, Control Peptide for AB1787P |
| Quantity | 100 μg |
| Regulatory Status | RUO - research use only |