missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Dopamine D2 Receptor Control Peptide

Catalog No. AG221MI
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AG221MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AG221MI Supplier MilliporeSigma™ Supplier No. AG221

Specifications

Gene ID (Entrez) NM_000795.2, NM_016574.2
Concentration 1 mg/mL
Content And Storage -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Format Liquid
Formulation Liquid in PBS, pH 7.5 containing 0.05% sodium azide.
Gene Symbol DRD2, D, D2R, D2DR
Product Type Dopamine D2 Receptor, control peptide for AB5084P
Quantity 100 μg
Regulatory Status RUO