missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Creatine Phosphokinase, Rabbit Skeletal Muscle, Protein

Catalog No. 238420mg
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
238420MG 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 238420MG Supplier MilliporeSigma™ Supplier No. 238420MG
Only null left
Native creatine phosphokinase from rabbit muscle. Catalyzes the conversion of creatine and ATP to creatine phosphate and ADP. Has an optimal pH of 9.0 for the forward reaction and 6.0-7.0 for the reverse reaction.

Specifications

Preparation Method 0.5% NaHCO3, pH 9.0 or 0.1M Imidazole Buffer, pH 6.6
Quantity 20 mg
Immunogen Creatine Phosphokinase
Storage Requirements Following reconstitution in 0.5% NaHCO3, refrigerate (4°C). Do not freeze or shake, as SH groups may be oxidized and form linkages between protein molecules, resulting in precipitation, turbidity, and loss of activity. NaHCO3 stock solutions are stable for 2-3 days at 4°C. Following reconstitution in 0.1 M imidazole buffer, aliquot and freeze (−20°C). Imidazole stock solutions are stable for up to 4 weeks at −20°C. Avoid freeze/thaw cycles, as 20-30% of activity will be lost with each freeze/thaw cycle.
Regulatory Status RUO
Species Rabbit
Recombinant Native
Form Lyophilized