missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ CpGenome™ Cytosine DNA Standard
Description
- Linear, double-stranded DNA with unmodified cytosines.
Specifications
Specifications
| Content And Storage | -20°C |
| Description | Cytosine DNA Standard |
| For Use With (Application) | Analyzing, quantifying cytosine modifications |
| Includes | One vial containing 2μg (40μL) of CpGenome™ Cytosine DNA Standard at a concentration of 50ng/μL. |
| Quantity | 2 μg/Vial |