Learn More
MilliporeSigma™ CpGenome™ 5-mC DNA Standard
For analyzing, quantifying cytosine modifications
Supplier:  MilliporeSigma™ S8005M
Catalog No. S8005M
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
-20°C
Analyzing, quantifying cytosine modifications
2 μg/Vial
5-mC DNA Standard
One vial containing 2μg (40μL) of CpGenome™ 5-mC DNA Standard at a concentration of 50ng/μL
Product Suggestions
Product Suggestions