missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ CpGenome™ 5-mC and 5-hmC DNA Standard Set
Description
- Sequence-matched DNA standards contain methylated, hydroxymethylated or unmodified cytosines.
Specifications
Specifications
| Description | 5-mC and 5-hmC DNA Standard Set |
| For Use With (Application) | Analyzing, quantifying cytosine modifications |
| Includes | Three vials, each containing 2μg (40μL) of CpGenome™ Cytosine DNA Standard, CpGenome™ 5-mC DNA Standard, or CpGenome™ 5-hmC DNA Standard, at a concentration of 50ng/μL |
| Quantity | 2 μg/Vial |