missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Complete Cell Media w/15% FBS Serum and LIF

Catalog No. ES101B
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ES101B 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ES101B Supplier MilliporeSigma™ Supplier No. ES101B

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ready to use medium qualified for mouse embryonic stem (ES) cell culture.

Ready to use medium formulated with 15% fetal bovine serum (FBS) and mouse leukemia inhibitory factor (LIF). Each lot is qualified for mouse embryonic stem (ES) cell culture.

Specifications

For Use With (Application) Cell culture and embryonic stem cells
Quantity 500 mL
Content And Storage 4°C up to 6 months