Learn More
MilliporeSigma™ Chromolith™ Column Coupler
Increase the separation efficiency by coupling several HPLC columns
Supplier:  MilliporeSigma™ 1.51467.0001
Catalog No. M1514670001
Only null left
Description
Description
  • With the column coupler it is possible to increase the plate count by coupling several columns in series
  • Coupling produces a column with a theoretical plate count significantly higher compared to particulate columns, while producing pressures still well below the HPLC system limit
Specifications
Specifications
Column Coupler
Product Suggestions
Product Suggestions