missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ TRAPeze™ XL Telomerase Detection Kit

Catalog No. S7707
Change view
Click to view available options
Quantity:
112 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7707 112 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7707 Supplier MilliporeSigma™ Supplier No. S7707

missing translation for 'validMainframeMsg'

For sensitive, quantitative measurement of telomerase activity in cell, tissue extracts using Amplifluor primer technology

  • Fluorometric detection format.
  • Includes primer modifications to reduce amplification artifacts and an internal PCR control.

Specifications

Description TRAPeze XL telomerase detection kit
Detection Method Fluorometric
For Use With (Application) PCR
Includes CHAPS lysis buffer, 5X TRAPEZE XL Reaction Mix, TS primer, RP Amplifluor primer, K2 Amplifluor primer, TSK2 template, dA, dC, dG and dTTP, Diluted in: 100mM Tris-HCl, pH 8.3, 7.5mM MgCl2,315mM KCl, 0.25% Tween 20, 5mM EGTA, 0.5 mg/mL BSA, PCR - Grade Water, protease, DNase, and RNase-free, deionized, TSR8, TSR8 template, Control cell pellet, telomerase positive cells
Quantity 112 Assays
Type Detection Assay Kit