missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ TRAPeze™ Telomerase Positive Control Cell Pellet

Catalog No. S7701
Change view
Click to view available options
Quantity:
1 Vial (1 x 106 Cells)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7701 1 Vial (1 x 106 Cells)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7701 Supplier MilliporeSigma™ Supplier No. S7701

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with components of TRAPEZE, TRAPEZE XL, TRAPEZE ELISA telomerase detection kit

Specifications

Content And Storage -75°C to -85deg;C
Description TRAPeze telomerase positive control cell pellet
Quantity 1 Vial (1 x 106 Cells)