missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ TRAPeze™ Telomerase Detection Kit

Catalog No. S7700
Change view
Click to view available options
Quantity:
112 Assays
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7700 112 Assays
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7700 Supplier MilliporeSigma™ Supplier No. S7700

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Detecting telomerase activity based on TRAP assay (Telomeric Repeat Amplification Protocol)

  • Includes an internal PCR control.
  • Primer modifications reduce amplification artifacts.

Specifications

Description TRAPeze telomerase detection kit
Detection Method Chromogenic
For Use With (Application) Detecting telomerase activity based on TRAP assay (Telomeric Repeat Amplification Protocol)
Includes The kit provides enough reagents to perform 112 TRAPEZE Kit assays with 50 ul reaction volume. A 25 ul reaction volume can also be used. With these reagents, 40 samples with appropriate positive and negative controls can be analyzed.
Quantity 112 Assays
Type Activity Assay Kit