missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ TRAPeze™ 1X CHAPS Lysis Buffer

Catalog No. S7705
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S7705 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S7705 Supplier MilliporeSigma™ Supplier No. S7705

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Used in extraction step of procedure

Specifications

Content And Storage 4°C
Description TRAPeze 1X CHAPS lysis buffer
Quantity 10 mL