missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ ReNcell NSC Maintenance Media

Catalog No. SCM005
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SCM005 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SCM005 Supplier MilliporeSigma™ Supplier No. SCM005

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

a defined serum-free, growth factor-free medium that has been optimized for the growth and in vitro differentiation of ReNcell immortalized human neural progenitor cells.

ReNcell NSC Maintenance Medium is a defined serum-free, growth factor-free medium that has been optimized for the growth and in vitro differentiation of ReNcell immortalized human neural progenitor cells. When used in conjunction with FGF and EGF, the maintenance medium will allow for the proliferation of ReNcell immortalized VM and CX neural stem cells. Withdrawal of the growth factors from ReNcell NSC Maintenance Medium will result in the spontaneous differentiation of ReNcell immortalized neural progenitor cells

Specifications

Classification Serum-free
For Use With (Application) Stem Cell Culture
Quantity 500 mL
Content And Storage -20°C