missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ QCM™ Tumor Cell Transendothelial Migration Assay

Catalog No. ECM558
Change view
Click to view available options
Quantity:
1 Kit (24 Assays)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM558 1 Kit (24 Assays)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM558 Supplier MilliporeSigma™ Supplier No. ECM558

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Analyzing ability of leukocytes to migrate through endothelium

Specifications

Content And Storage TNFalpha should be stored at -20°C. All other components should be stored at 2-8°C
Description QCM Tumor Cell Transendothelial Migration Assay
Detection Method Colorimetric
For Use With (Application) Analyzing ability of leukocytes to migrate through endothelium
Includes Migration test plate, TNFalpha, cell stain solution, extraction buffer, 24-, 96-well stain extraction/quantitation plate, swabs, forceps
Quantity 1 Kit (24 Assays)