missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ QCM™ Leukocyte Transendothelial Migration Assay

Catalog No. ECM557
Change view
Click to view available options
Quantity:
1 Kit (24 Assays)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM557 1 Kit (24 Assays)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM557 Supplier MilliporeSigma™ Supplier No. ECM557

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For analyzing ability of leukocytes to migrate through endothelium

Specifications

Description QCM Leukocyte Transendothelial Migration Assay
Detection Method Colorimetric
For Use With (Application) Analyzing ability of leukocytes to migrate through endothelium
Includes Migration test plate, TNFalpha, cell stain solution, WST-1 reagent, electro coupling solution, 96-well stain quantitation plate, forceps
Quantity 1 Kit (24 Assays)