Learn More
MilliporeSigma™ Chemicon™ QCM Laminin Migration Assay, 24-well, Fluorimetric
Includes 5μm porous membrane Boyden chamber and mouse laminin, major extracellular matrix
Supplier:  MilliporeSigma™ ECM221
Catalog No. ECM221
Only null left
Description
Description
Specifications
Specifications
Laminin Migration Assay
Fluorescence
Calcein-AM, 24-well Cell Migration Plate Assembly, BSA 30% solution, 24-well Stain Receiver Plate, 96-Well Black Quantitation Plate, Forceps; Laminin; Accutase
1/Pk.
Product Suggestions
Product Suggestions