missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ QCM™ Haptotaxis Cell Migration Assay-Fibronectin

Catalog No. ECM580
Change view
Click to view available options
Quantity:
1 Kit (Sufficient for analysis of 12 samples)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM580 1 Kit (Sufficient for analysis of 12 samples)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM580 Supplier MilliporeSigma™ Supplier No. ECM580

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Study of epithelial and fibroblast cell haptotaxis

Specifications

Content And Storage 4°C for up to one year 
Description QCM Haptotaxis Cell Migration Assay-Fibronectin
Detection Method Colorimetric
For Use With (Application) Study of epithelial and fibroblast cell haptotaxis
Includes Fibronectin test, BSA control, 24-, 96-well stain extraction/quantitation plate, cell stain solution, extraction buffer, swabs, forceps
Quantity 1 Kit (Sufficient for analysis of 12 samples)