missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ QCM™ Collagen Cell Invasion Assay, 24-well (8μm), Fluorimetric
Description
- Fluorometric detection format.
Specifications
Specifications
| Content And Storage | 2°C to 8°C |
| Description | QCM Collagen Cell Invasion Assay kit |
| Detection Method | Fluorescence |
| For Use With (Application) | For evaluation of invasive tumor cells |
| Quantity | 1/Pk. |
| Type | Cell Invasion Assay |