missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ QCM™ Collagen Cell Invasion Assay

Catalog No. ECM556
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM556 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM556 Supplier MilliporeSigma™ Supplier No. ECM556

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Evaluation of invasive tumor cells

Specifications

Content And Storage 2°C to 8°C
Description QCM Collagen Cell Invasion Assay, 96-well (8μm)
Detection Method Fluorescence
For Use With (Application) Evaluation of invasive tumor cells
Includes Sterile 96-well plate assembly, cell detachment solution, 4X cell lysis buffer, CyQuant GR Dye 1, 96-well cell culture tray
Quantity 1 Plate