missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ QCM Chemotaxis Cell Migration Assay, 96-well (5μm), Fluorimetric
For studying monocyte/macrophage migration
Supplier: MilliporeSigma™ ECM512
Specifications
| QCM Chemotaxis Cell Migration Assay | |
| Fluorescence | |
| Activity Assay | |
| 1/Pk. |