missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ QCM Chemotaxis Cell Migration Assay, 96-well (3μm), Fluorimetric

Catalog No. ECM515
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM515 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM515 Supplier MilliporeSigma™ Supplier No. ECM515

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activity Assay

  • Fluorometric detection format.
  • Includes cell detachment solution, lysis buffer and CyQuant GR dye.

Specifications

Description QCM Chemotaxis Cell Migration Assay
Detection Method Fluorescence
For Use With (Application) Activity Assay
Includes Forceps, Sterile 3μm 96well Cell migration plate, 96 well culture tray, Cell detachment soln., 4X cell lysis buffer, CyQUANT GR Dye
Quantity 1/Pk.
Type Cell-Based Assay Kit
Druggable Target Cell-based Assay