missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ QCM Chemotaxis Cell Migration Assay, 24-well (8μm), Fluorimetric

Catalog No. ECM509
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ECM509 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ECM509 Supplier MilliporeSigma™ Supplier No. ECM509

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activity Assay

  • Fluorometric detection format.
  • Suitable for quantitative screening of pharmacological agents.

Specifications

Content And Storage 2°C to 8°C
Description QCM Chemotaxis Cell Migration Assay
Detection Method Fluorescence
For Use With (Application) Activity Assay
Includes Forceps, Sterile 8μm 24-well Cell migration plate, two 24-well plates w/12 inserts/plate, Cell detachment soln., 4X cell lysis buffer, CyQUANT GR Dye
Quantity 1/Pk.
Type Cell-Based Assay Kit
Druggable Target Cell-based Assay